This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYB5B SIRNA Assays ›
Gene Symbol:
CYB5B
Gene Name:
cytochrome b5 type B
Gene Aliases:
CYB5-M, CYPB5M, OMB5
Chromosome Location:
Chr.16: 69424595 - 69466264 on Build GRCh38
UniGene ID:
Hs.461131
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s37387

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CYB5B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030579.2 NP_085056.2 2 327
GenBank mRNA AB009282.1 - 2 354
AK291576.1 - 2 259
BC004373.1 - 2 262 3639133
BC014431.1 - 2 285 3542294
BX647922.1 - 2 303

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
protein binding
heme binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header