This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSGA10 SIRNA Assays ›
Gene Symbol:
TSGA10
Gene Name:
testis specific 10
Gene Aliases:
CEP4L, CT79
Chromosome Location:
Chr.2: 98997261 - 99154962 on Build GRCh38
UniGene ID:
Hs.120267
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37299
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
357.00
Your Price
Online offer:
296.31
296.3
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSGA10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025244.2
NP_079520.1
14
1648
NM_182911.3
NP_878915.2
13
1627
XM_005264023.1
XP_005264080.1
13
1602
XM_005264024.2
XP_005264081.1
15
1767
XM_005264025.1
XP_005264082.1
14
1738
XM_005264026.1
XP_005264083.1
15
1723
XM_005264027.1
XP_005264084.1
13
1558
XM_005264029.1
XP_005264086.1
13
1547
XM_006712781.2
XP_006712844.1
16
1801
XM_011511934.1
XP_011510236.1
13
1637
XM_011511935.2
XP_011510237.1
12
1411
XM_011511937.1
XP_011510239.1
13
1608
XM_011511939.1
XP_011510241.1
16
1896
XM_011511940.2
XP_011510242.1
15
1867
XM_011511941.1
XP_011510243.1
16
1940
XM_011511942.1
XP_011510244.1
16
1930
XM_011511943.2
XP_011510245.1
12
1378
XM_017005035.1
XP_016860524.1
13
1628
XM_017005036.1
XP_016860525.1
13
1637
XM_017005037.1
XP_016860526.1
13
1637
XM_017005038.1
XP_016860527.1
13
1637
XM_017005039.1
XP_016860528.1
16
1790
XM_017005040.1
XP_016860529.1
15
1891
XM_017005041.1
XP_016860530.1
13
1749
XM_017005042.1
XP_016860531.1
15
1779
XM_017005043.1
XP_016860532.1
13
1612
XM_017005044.1
XP_016860533.1
15
1755
XM_017005045.1
XP_016860534.1
16
3942
XM_017005046.1
XP_016860535.1
15
1757
XM_017005047.1
XP_016860536.1
12
1665
XM_017005048.1
XP_016860537.1
12
1461
GenBank mRNA
AF254756.1
-
12
1623
AK304328.1
-
12
1491
AY014284.1
-
12
1476
BC028366.1
-
13
1628
4830604
BX647157.1
-
14
1841
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
cell projection assembly
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0