This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM134 SIRNA Assays ›
Gene Symbol:
TMEM134
Gene Name:
transmembrane protein 134
Gene Aliases:
-
Chromosome Location:
Chr.11: 67464348 - 67469277 on Build GRCh38
UniGene ID:
Hs.288761
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s37029

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM134
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078650.2 NP_001072118.1 3, 4 422
NM_001078651.2 NP_001072119.1 3, 4 395
NM_025124.3 NP_079400.1 3, 4 422
NR_073409.1 3, 4 422
NR_073410.1 3, 4 406
NR_073411.1 3, 4 433
NR_073412.1 3, 4 422
NR_073413.1 3, 4 406
XM_006718693.2 XP_006718756.1 3, 4 390
XM_011545268.2 XP_011543570.1 3, 4 417
XM_017018356.1 XP_016873845.1 3, 4 417
GenBank mRNA AK025402.1 - 3, 4 438
AK296988.1 - 3, 4 351
AK303342.1 - 3, 4 386
AK310743.1 - 3, 4 310
AY007143.1 - 3, 4 392
BC013883.1 - 3, 4 391 3678954
BC125134.1 - 3, 4 390 40119367
BG488634.1 - 3, 4 344
BG719036.1 - 3, 4 402
BM920758.1 - 390
BX400047.2 - 3, 4 378
CN336445.1 - 3, 4 256
DN999439.1 - 3, 4 411
HY053672.1 - 3, 4 422
HY059123.1 - 3, 4 349

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0