This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FRMD1 SIRNA Assays ›
Gene Symbol:
FRMD1
Gene Name:
FERM domain containing 1
Gene Aliases:
bA164L23.1
Chromosome Location:
Chr.6: 168055745 - 168093007 on Build GRCh38
UniGene ID:
Hs.266746
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s36765

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FRMD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122841.2 NP_001116313.1 4 355
NM_024919.4 NP_079195.3 4 505
NR_110312.1 5 883
XM_011536137.1 XP_011534439.1 5 585
XM_011536138.1 XP_011534440.1 5 401
XM_011536140.1 XP_011534442.1 4 440
XM_011536141.1 XP_011534443.1 4 380
XM_011536142.2 XP_011534444.1 4 418
XM_011536143.1 XP_011534445.1 4 316
XM_011536145.1 XP_011534447.1 4 355
XM_017011317.1 XP_016866806.1 5 585
GenBank mRNA AK074110.1 - 5 442
AK097179.1 - 4 505
AK097579.1 - 4 355
AK125963.1 - 5 855
AK303194.1 - 3 159
AK303407.1 - 2 213
AK316478.1 - 3 159
BC137144.1 - 4 463 9021141

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm