This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DOK3 SIRNA Assays ›
Gene Symbol:
DOK3
Gene Name:
docking protein 3
Gene Aliases:
DOKL
Chromosome Location:
Chr.5: 177501904 - 177510507 on Build GRCh38
UniGene ID:
Hs.720849
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s36666

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DOK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308235.1 NP_001295164.1 6 1905
NM_001308236.1 NP_001295165.1 6 1728
NM_024872.2 NP_079148.2 6 1684
XM_005265985.3 XP_005266042.1 6 1741
XM_011534654.2 XP_011532956.1 6 1618
XM_011534655.2 XP_011532957.1 6 1794
XM_011534656.2 XP_011532958.1 6 1595
XM_011534657.2 XP_011532959.1 5 1445
XM_017009883.1 XP_016865372.1 4 1416
GenBank mRNA AK026223.1 - 6 1679

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
insulin receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0