This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSTCD SIRNA Assays ›
Gene Symbol:
GSTCD
Gene Name:
glutathione S-transferase C-terminal domain containing
Gene Aliases:
-
Chromosome Location:
Chr.4: 105708778 - 105848023 on Build GRCh38
UniGene ID:
Hs.161429
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s36377

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GSTCD
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031720.3 NP_001026890.2 3 781
NM_024751.3 NP_079027.2 3 702
XM_005263222.3 XP_005263279.1 3 969
XM_011532248.2 XP_011530550.1 3 925
XM_011532249.2 XP_011530551.1 3 787
XM_011532252.2 XP_011530554.1 3 1159
GenBank mRNA AK023335.1 - 3 960
AK225563.1 - 3 960
AK292355.1 - 3 784
BC032942.2 - 3 781 4838672
BX648355.1 - 3 703

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0