This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF128 SIRNA Assays ›
Gene Symbol:
RNF128
Gene Name:
ring finger protein 128, E3 ubiquitin protein ligase
Gene Aliases:
GRAIL
Chromosome Location:
Chr.X: 106693838 - 106797016 on Build GRCh38
UniGene ID:
Hs.496542
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35869

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF128
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024539.3 NP_078815.3 7 1291
NM_194463.1 NP_919445.1 7 1454
GenBank mRNA AF394689.1 - 7 1214
AK027169.1 - 5 463
AK074264.1 - 7 1454
AK126553.1 - 7 1274
BC030951.1 - 6 567
BC036901.1 - 7 1303
BC056677.1 - 5 438
BC063404.1 - 7 1293 5191785

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of protein stability
negative regulation of cytokine biosynthetic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein localization to lysosome
positive regulation of protein catabolic process in the vacuole
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header