This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF343 SIRNA Assays ›
Gene Symbol:
ZNF343
Gene Name:
zinc finger protein 343
Gene Aliases:
dJ734P14.5
Chromosome Location:
Chr.20: 2481817 - 2524522 on Build GRCh38
UniGene ID:
Hs.516846
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35684

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF343
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282495.1 NP_001269424.1 7 899
NM_001282496.1 NP_001269425.1 6 931
NM_001282497.1 NP_001269426.1 7 1096
NM_001282498.1 NP_001269427.1 4 647
NM_001321800.1 NP_001308729.1 6 837
NM_001321801.1 NP_001308730.1 7 960
NM_001321802.1 NP_001308731.1 7 1093
NM_001321803.1 NP_001308732.1 7 957
NM_001321805.1 NP_001308734.1 6 970
NM_024325.5 NP_077301.4 6 973
XM_017028062.1 XP_016883551.1 6 814
GenBank mRNA AK055734.1 - 2 402
AK096911.1 - 6 974
BC011862.2 - 1 143
BC065009.1 - 7 896 6166526
DA034001.1 - 4 478

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0