This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNDC11 SIRNA Assays ›
Gene Symbol:
FNDC11
Gene Name:
fibronectin type III domain containing 11
Gene Aliases:
C20orf195
Chromosome Location:
Chr.20: 63553020 - 63556712 on Build GRCh38
UniGene ID:
Hs.197755
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35442

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FNDC11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319152.1 NP_001306081.1 2 908
NM_001319153.1 NP_001306082.1 2 910
NM_024059.3 NP_076964.1 2 926
XM_011529041.2 XP_011527343.1 2 1463
XM_017028060.1 XP_016883549.1 2 1062
GenBank mRNA AK125168.1 - 1 2783
BC000912.2 - 2 926 3048411

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0