This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP4F12 SIRNA Assays ›
Gene Symbol:
CYP4F12
Gene Name:
cytochrome P450 family 4 subfamily F member 12
Gene Aliases:
CYPIVF12, F22329_1
Chromosome Location:
Chr.19: 15673018 - 15698819 on Build GRCh38
UniGene ID:
Hs.131459
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s35343

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CYP4F12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023944.3 NP_076433.3 10 1339
NR_117085.1 9 1357
XM_006722850.3 XP_006722913.1 10 1224
XM_011528202.2 XP_011526504.1 8 859
XM_011528203.2 XP_011526505.1 7 2081
XM_011528204.2 XP_011526506.1 8 1560
XM_011528205.2 XP_011526507.1 7 772
XM_011528207.2 XP_011526509.1 7 2468
XM_011528208.2 XP_011526510.1 9 978
GenBank mRNA AB035130.1 - 10 1250
AB035131.1 - 10 1250
AK074412.1 - 4 2641
AK075435.1 - 9 1285
AL832171.1 - 9 2246
AY008841.1 - 9 1220
AY358977.1 - 10 1261
BC035350.1 - 10 1266 5187264

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
long-chain fatty acid metabolic process
renal water homeostasis
pressure natriuresis
drug metabolic process
arachidonic acid metabolic process
epoxygenase P450 pathway
leukotriene B4 catabolic process
vitamin E metabolic process
sodium ion homeostasis
oxidation-reduction process
iron ion binding
arachidonic acid epoxygenase activity
alkane 1-monooxygenase activity
oxygen binding
heme binding
leukotriene-B4 20-monooxygenase activity
aromatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header