This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNDC3B SIRNA Assays ›
Gene Symbol:
FNDC3B
Gene Name:
fibronectin type III domain containing 3B
Gene Aliases:
FAD104, PRO4979, YVTM2421
Chromosome Location:
Chr.3: 172039628 - 172400703 on Build GRCh38
UniGene ID:
Hs.744888
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s34922

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FNDC3B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135095.1 NP_001128567.1 3 243
NM_022763.3 NP_073600.3 3 316
XM_017007062.1 XP_016862551.1 3 296
XM_017007064.1 XP_016862553.1 3 296
GenBank mRNA AB098597.1 - 2 201
AK127826.1 - 3 296
AK223599.1 - 3 347
AK314478.1 - 3 243
AY358367.1 - 3 285
BC012204.1 - 3 314
BC039297.1 - 3 207 5297602
BX648340.1 - 3 238

Back To Top

More Information


Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0