This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTC31 SIRNA Assays ›
Gene Symbol:
TTC31
Gene Name:
tetratricopeptide repeat domain 31
Gene Aliases:
-
Chromosome Location:
Chr.2: 74483032 - 74494564 on Build GRCh38
UniGene ID:
Hs.557709
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s34743

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TTC31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022492.4 NP_071937.4 8 724
NR_027749.1 7 621
XM_005264492.1 XP_005264549.1 8 713
XM_011533035.1 XP_011531337.1 8 713
XM_011533036.1 XP_011531338.1 8 713
XM_011533037.1 XP_011531339.1 8 713
XM_011533040.1 XP_011531342.1 7 662
XM_017004706.1 XP_016860195.1 8 713
XM_017004707.1 XP_016860196.1 8 713
XM_017004708.1 XP_016860197.1 7 618
XM_017004709.1 XP_016860198.1 7 848
GenBank mRNA AK022850.1 - 8 710
AK090520.1 - 7 763
AK223355.1 - 8 708
AK303105.1 - 7 607
BC036017.1 - 8 708 5270575
BC045536.1 - 7 612
BC096709.1 - 8 710 40023059
BC098365.1 - 8 710 40023060
BC099729.1 - 6 995
BX648131.1 - 5 1176

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0