This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NYX SIRNA Assays ›
Gene Symbol:
NYX
Gene Name:
nyctalopin
Gene Aliases:
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
Chromosome Location:
Chr.X: 41447460 - 41476414 on Build GRCh38
UniGene ID:
Hs.302019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s34115

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NYX
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022567.2 NP_072089.1 2 2577
XM_017029709.1 XP_016885198.1 3 2245
GenBank mRNA AF254868.1 - 3 2245
AJ278865.1 - 2 2603

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
visual perception
biological_process
cytokine-mediated signaling pathway
negative regulation of JAK-STAT cascade
molecular_function
protein kinase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0