This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIAA1524 SIRNA Assays ›
Gene Symbol:
KIAA1524
Gene Name:
KIAA1524
Gene Aliases:
CIP2A, p90
Chromosome Location:
Chr.3: 108545752 - 108589644 on Build GRCh38
UniGene ID:
Hs.591308
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s33522

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIAA1524
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020890.2 NP_065941.2 11 1611
XM_006713716.3 XP_006713779.1 11 1395
XM_006713717.2 XP_006713780.1 12 1507
XM_011513056.1 XP_011511358.1 11 1355
XM_011513057.2 XP_011511359.1 12 1476
GenBank mRNA AB040957.1 - 11 1338
AF334474.1 - 11 1410
AK022912.1 - 2 222
AK056933.1 - 11 1420
AK308315.1 - 11 1357
AK310446.1 - 11 1615
AL713689.1 - 11 1406
BC063433.1 - 11 1402
BC082961.1 - 11 1364
BC130564.1 - 11 1378 40146595
BC136371.1 - 11 1378 9020358

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
cell-cell adhesion
positive regulation of neural precursor cell proliferation
protein binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm