This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIAA1462 SIRNA Assays ›
Gene Symbol:
KIAA1462
Gene Name:
KIAA1462
Gene Aliases:
JCAD
Chromosome Location:
Chr.10: 30012800 - 30069816 on Build GRCh38
UniGene ID:
Hs.533953
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s33429

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIAA1462
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020848.2 NP_065899.1 3 2385
XM_011519608.1 XP_011517910.1 3 2424
XM_011519609.2 XP_011517911.1 4 2452
GenBank mRNA AB040895.2 - 2 2278
AK308850.1 - 3 2367
AL583916.1 - 1 322
BX648192.1 - 4 2474
BX648263.1 - 1 1309
BX648773.1 - 1 1435

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cell adhesion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0