This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COG6 SIRNA Assays ›
Gene Symbol:
COG6
Gene Name:
component of oligomeric golgi complex 6
Gene Aliases:
CDG2L, COD2, SHNS
Chromosome Location:
Chr.13: 39655627 - 39791665 on Build GRCh38
UniGene ID:
Hs.507805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s33195

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: COG6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145079.1 NP_001138551.1 11, 12 1173
NM_020751.2 NP_065802.1 11, 12 1173
NR_026745.1 12, 13 1238
XM_011535168.1 XP_011533470.1 11, 12 1123
GenBank mRNA AB032960.1 - 11, 12 1063
AF116827.1 - 12, 13 1121
AK026638.1 - 10, 11 840
AK294443.1 - 11, 12 1173
BC051723.1 - 11, 12 1123 5263056
CR627406.1 - 11, 12 1116

Back To Top

More Information


Gene Ontology Categories:

Function(s)

ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
protein transport
glycosylation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0