This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PARL SIRNA Assays ›
Gene Symbol:
PARL
Gene Name:
presenilin associated rhomboid like
Gene Aliases:
PRO2207, PSARL, PSARL1, PSENIP2, RHBDS1
Chromosome Location:
Chr.3: 183826488 - 183884908 on Build GRCh38
UniGene ID:
Hs.478469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30888

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PARL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037639.2 NP_001032728.1 2 247
NM_001324436.1 NP_001311365.1 2 247
NM_001324437.1 NP_001311366.1 2 247
NM_001324438.1 NP_001311367.1 2 247
NM_018622.6 NP_061092.3 2 247
NR_136893.1 2 247
XM_005247582.4 XP_005247639.1 2 242
XM_017006800.1 XP_016862289.1 2 242
XM_017006801.1 XP_016862290.1 2 240
XM_017006802.1 XP_016862291.1 2 240
XM_017006803.1 XP_016862292.1 2 238
GenBank mRNA AF197937.1 - 2 185
AK024765.1 - 2 219
BC003653.1 - 2 234 3537438
BC014058.2 - 2 205 3937238
BG714761.1 - 2 245
CB122278.1 - 2 244

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001005767, AK179606, AK207637, BC083153, AK151652, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein processing
regulation of proteolysis
membrane protein proteolysis
negative regulation of release of cytochrome c from mitochondria
regulation of mitophagy
regulation of protein targeting to mitochondrion
regulation of reactive oxygen species metabolic process
negative regulation of intrinsic apoptotic signaling pathway
endopeptidase activity
serine-type endopeptidase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0