This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM144 SIRNA Assays ›
Gene Symbol:
TMEM144
Gene Name:
transmembrane protein 144
Gene Aliases:
-
Chromosome Location:
Chr.4: 158201597 - 158255287 on Build GRCh38
UniGene ID:
Hs.176227
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30706

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM144
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018342.4 NP_060812.2 10 1298
XM_005263110.1 XP_005263167.1 11 1253
XM_005263112.3 XP_005263169.1 12 1288
XM_006714254.1 XP_006714317.1 11 1110
XM_017008366.1 XP_016863855.1 9 1266
XM_017008367.1 XP_016863856.1 8 838
XM_017008368.1 XP_016863857.1 9 998
GenBank mRNA AK002017.1 - 9 1202
BC026318.1 - 9 924
BC054487.1 - 10 1046 5732836
BC060875.1 - 10 1024 4800219

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0