siRNA Details
Gene Symbol: PRR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018280.2 | NP_060750.1 | 2 | 589 | ||
GenBank mRNA | AK001807.1 | - | 2 | 589 | ||
AK096201.1 | - | 2 | 241 |

Gene Symbol: PRR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018280.2 | NP_060750.1 | 2 | 589 | ||
GenBank mRNA | AK001807.1 | - | 2 | 589 | ||
AK096201.1 | - | 2 | 241 |