This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC20 SIRNA Assays ›
Gene Symbol:
LRRC20
Gene Name:
leucine rich repeat containing 20
Gene Aliases:
-
Chromosome Location:
Chr.10: 70298970 - 70382650 on Build GRCh38
UniGene ID:
Hs.7778
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30479

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278211.1 NP_001265140.1 5 918
NM_001278212.1 NP_001265141.1 5 863
NM_001278213.1 NP_001265142.1 4 662
NM_001278214.1 NP_001265143.1 4 718
NM_018205.3 NP_060675.1 4 695
NM_018239.3 NP_060709.2 4 713
NM_207119.2 NP_997002.1 5 1176
NR_103467.1 3 568
NR_103468.1 3 550
XM_017016374.1 XP_016871863.1 5 858
XM_017016375.1 XP_016871864.1 5 853
GenBank mRNA AK001613.1 - 4 671
AK001706.1 - 4 689
AK094734.1 - 5 859
AL122117.1 - 2 400
AY358318.1 - 5 920
BC024001.2 - 5 854 4822655
CF552977.1 - 5 835
DA032832.1 - 3 542
DA386385.1 - 3 524

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0