This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FANCI SIRNA Assays ›
Gene Symbol:
FANCI
Gene Name:
Fanconi anemia complementation group I
Gene Aliases:
KIAA1794
Chromosome Location:
Chr.15: 89243954 - 89317132 on Build GRCh38
UniGene ID:
Hs.513126
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30461

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FANCI
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113378.1 NP_001106849.1 13 1351
NM_018193.2 NP_060663.2 13 1351
XM_011521756.2 XP_011520058.1 13 1375
XM_011521757.2 XP_011520059.1 13 1371
XM_011521764.2 XP_011520066.1 13 1375
XM_011521767.2 XP_011520069.1 13 1345
GenBank mRNA AK001581.1 - 7 740
AK027564.1 - 2 238
AK055176.1 - 13 1310
BC140769.1 - 13 1308 9021637
BC144483.1 - 13 1308 9053010
EF469766.1 - 12 1261
EF567077.1 - 12 1261

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic G2 DNA damage checkpoint
positive regulation of protein ubiquitination
interstrand cross-link repair
DNA binding
protein binding
DNA polymerase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0