This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NUDT11 SIRNA Assays ›
Gene Symbol:
NUDT11
Gene Name:
nudix hydrolase 11
Gene Aliases:
APS1, ASP1, DIPP3b, DIPP3beta, hDIPP3beta
Chromosome Location:
Chr.X: 51490011 - 51496607 on Build GRCh38
UniGene ID:
Hs.200016
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30391

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NUDT11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018159.3 NP_060629.2 2 1291
GenBank mRNA AK001490.1 - 2 1178
BC009942.2 - 2 1245 4303422

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

inositol phosphate metabolic process
diphosphoinositol-polyphosphate diphosphatase activity
metal ion binding
inositol diphosphate tetrakisphosphate diphosphatase activity
inositol bisdiphosphate tetrakisphosphate diphosphatase activity
inositol diphosphate pentakisphosphate diphosphatase activity
inositol-1-diphosphate-2,3,4,5,6-pentakisphosphate diphosphatase activity
inositol-3-diphosphate-1,2,4,5,6-pentakisphosphate diphosphatase activity
inositol-5-diphosphate-1,2,3,4,6-pentakisphosphate diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 1-diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
inositol-3,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0