This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MSL2 SIRNA Assays ›
Gene Symbol:
MSL2
Gene Name:
male-specific lethal 2 homolog (Drosophila)
Gene Aliases:
MSL-2, MSL2L1, RNF184
Chromosome Location:
Chr.3: 136148918 - 136197300 on Build GRCh38
UniGene ID:
Hs.18631
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s30332

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MSL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145417.1 NP_001138889.1 2 230
NM_018133.3 NP_060603.2 2 1031
XM_005247571.3 XP_005247628.1 2 765
XM_006713684.3 XP_006713747.1 2 248
XM_011512949.2 XP_011511251.1 2 335
GenBank mRNA AB046805.1 - 2 379
AK091956.1 - 2 1031
AK292557.1 - 2 526
AK302491.1 - 2 197
AL834289.1 - 2 349
BC093764.1 - 2 331 7939609
BC093790.1 - 2 330 7939635
DC409643.1 - 2 316

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
histone H4-K16 acetylation
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0