This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD9 SIRNA Assays ›
Gene Symbol:
KCTD9
Gene Name:
potassium channel tetramerization domain containing 9
Gene Aliases:
BTBD27
Chromosome Location:
Chr.8: 25424357 - 25458468 on Build GRCh38
UniGene ID:
Hs.188763
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29445

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KCTD9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017634.3 NP_060104.2 4 519
GenBank mRNA AK298518.1 - 4 323
BC021216.2 - 4 322 4302317
BC068518.1 - 4 332 5265435
BG249272.1 - 453
BU149129.1 - 4 437
DA115324.1 - 4 520

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein homooligomerization
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0