This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MXRA8 SIRNA Assays ›
Gene Symbol:
MXRA8
Gene Name:
matrix remodeling associated 8
Gene Aliases:
-
Chromosome Location:
Chr.1: 1352689 - 1363541 on Build GRCh38
UniGene ID:
Hs.515687
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29242

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MXRA8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282582.1 NP_001269511.1 5 483
NM_001282583.1 NP_001269512.1 4 902
NM_001282584.1 NP_001269513.1 3 402
NM_001282585.1 NP_001269514.1 4 458
NM_032348.3 NP_115724.1 4 705
XM_017001516.1 XP_016857005.1 4 482
XM_017001517.1 XP_016857006.1 3 2109
GenBank mRNA AB052096.1 - 5 483
AK027269.1 - 4 458
AK095966.1 - 4 902
AK225028.1 - 4 444
AK293442.1 - 3 171
AK304615.1 - 4 442
AK315510.1 - 4 472
AY358391.1 - 3 148
BC006213.2 - 4 446 3343261
BC017312.2 - 4 446 5090754

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024263, AK004732, BC026438, EU849596, EU849597, and others.

Gene Ontology Categories:

Function(s) Process(es)

establishment of glial blood-brain barrier
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0