This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRMT13 SIRNA Assays ›
Gene Symbol:
TRMT13
Gene Name:
tRNA methyltransferase 13 homolog
Gene Aliases:
CCDC76
Chromosome Location:
Chr.1: 100133080 - 100150498 on Build GRCh38
UniGene ID:
Hs.729095
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29058

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRMT13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019083.2 NP_061956.2 1, 2 174
NR_135078.1 1, 2 174
XM_005270945.3 XP_005271002.1 1, 2 244
XM_005270946.2 XP_005271003.1 1, 2 164
XM_017001504.1 XP_016856993.1 1, 2 161
XM_017001505.1 XP_016856994.1 1, 2 192
GenBank mRNA AK001149.1 - 1, 2 159
AK002081.1 - 1, 2 161
AK289404.1 - 1, 2 161
AK294738.1 - 1, 2 159
AK295828.1 - 1, 2 171
AK298941.1 - 1, 2 159
AL699548.1 - 1, 2 175
BC057226.1 - 1, 2 174
BC075811.1 - 1, 2 171 5104751

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

tRNA methylation
tRNA methyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0