This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM64A SIRNA Assays ›
Gene Symbol:
FAM64A
Gene Name:
family with sequence similarity 64 member A
Gene Aliases:
CATS, RCS1
Chromosome Location:
Chr.17: 6444305 - 6451065 on Build GRCh38
UniGene ID:
Hs.592116
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s29052

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM64A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195228.1 NP_001182157.1 2 377
NM_019013.2 NP_061886.2 2 377
XM_011523944.2 XP_011522246.1 2 487
GenBank mRNA AK001018.1 - 1 317
AK001353.1 - 2 337
AK225613.1 - 2 339
AK225702.1 - 2 337
BC005004.1 - 2 344 3533875
BC013966.2 - 2 323 2988318
KU178653.1 - 1 268
KU178654.1 - 1 268

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic nuclear division
cell division
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0