This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C14ORF166 SIRNA Assays ›
Gene Symbol:
C14orf166
Gene Name:
chromosome 14 open reading frame 166
Gene Aliases:
CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1
Chromosome Location:
Chr.14: 51989510 - 52004702 on Build GRCh38
UniGene ID:
Hs.534457
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s28444

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C14orf166
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016039.2 NP_057123.1 2 251
GenBank mRNA AF087877.1 - 2 282
AF100755.1 - 2 282
AF151857.1 - 2 282
AK312004.1 - 2 215
BC001722.1 - 2 203 3528725
DQ525179.1 - 2 286

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
tRNA splicing, via endonucleolytic cleavage and ligation
viral process
positive regulation of transcription from RNA polymerase II promoter
RNA transport
RNA polymerase II core binding
RNA binding
protein binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0