This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BFAR SIRNA Assays ›
Gene Symbol:
BFAR
Gene Name:
bifunctional apoptosis regulator
Gene Aliases:
BAR, RNF47
Chromosome Location:
Chr.16: 14632811 - 14669236 on Build GRCh38
UniGene ID:
Hs.435556
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27840

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BFAR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016561.2 NP_057645.1 5 1015
XM_005255350.2 XP_005255407.1 4 790
XM_011522520.1 XP_011520822.1 3 620
GenBank mRNA AF173003.1 - 5 887
AK291114.1 - 5 823
AK297067.1 - 5 576
AK300778.1 - 3 588
AK313890.1 - 4 823
BC003054.1 - 5 846 3537783

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
apoptotic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
protein autoubiquitination
protein K63-linked ubiquitination
protein K48-linked ubiquitination
negative regulation of IRE1-mediated unfolded protein response
protein binding
zinc ion binding
protein binding, bridging
ubiquitin protein ligase activity
caspase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0