This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SIDT2 SIRNA Assays ›
Gene Symbol:
SIDT2
Gene Name:
SID1 transmembrane family member 2
Gene Aliases:
CGI-40
Chromosome Location:
Chr.11: 117179223 - 117197445 on Build GRCh38
UniGene ID:
Hs.712144
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27405

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SIDT2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040455.1 NP_001035545.1 6 707
GenBank mRNA AF151799.1 - 6 749
AK292501.1 - 6 1187
AK292974.1 - 6 698
AK303023.1 - 6 663
AY358442.1 - 6 716
BC114522.1 - 6 736 40035836
KC109742.1 - 6 663

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
type B pancreatic cell development
response to glucose
dsRNA transport
glucose homeostasis
type B pancreatic cell proliferation
regulation of insulin secretion involved in cellular response to glucose stimulus
RNA transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0