This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD207 SIRNA Assays ›
Gene Symbol:
CD207
Gene Name:
CD207 molecule
Gene Aliases:
CLEC4K
Chromosome Location:
Chr.2: 70825248 - 70860787 on Build GRCh38
UniGene ID:
Hs.199731
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s27008

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD207
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015717.4 NP_056532.4 3 600
XM_011532874.1 XP_011531176.1 4 788
XM_011532875.2 XP_011531177.1 5 805
XM_011532876.2 XP_011531178.1 5 804
GenBank mRNA AJ242859.1 - 3 600
AK314927.1 - 3 594
BC022278.1 - 3 593 4692155

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
receptor-mediated endocytosis
defense response to virus
mannose binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0