This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM216A SIRNA Assays ›
Gene Symbol:
FAM216A
Gene Name:
family with sequence similarity 216 member A
Gene Aliases:
C12orf24, HSU79274
Chromosome Location:
Chr.12: 110468424 - 110490387 on Build GRCh38
UniGene ID:
Hs.436618
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s26674

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM216A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013300.2 NP_037432.2 5 946
XM_005253875.4 XP_005253932.1 5 456
XM_011538246.2 XP_011536548.1 5 474
GenBank mRNA AK225731.1 - 5 937
AK296280.1 - 5 958
AK297684.1 - 5 456
AK308802.1 - 5 559
AK309603.1 - 4 470
BC020967.1 - 5 762 3848037
CR457001.1 - 5 497
U79274.1 - 5 913

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0