This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD22 SIRNA Assays ›
Gene Symbol:
CD22
Gene Name:
CD22 molecule
Gene Aliases:
SIGLEC-2, SIGLEC2
Chromosome Location:
Chr.19: 35329166 - 35347361 on Build GRCh38
UniGene ID:
Hs.579691
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s2613

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185099.1 NP_001172028.1 5 956
NM_001185100.1 NP_001172029.1 5 956
NM_001278417.1 NP_001265346.1 4 784
NM_001771.3 NP_001762.2 5 956
GenBank mRNA AK225625.1 - 5 938
AK225694.1 - 5 938
AK301177.1 - 5 819
BC109306.1 - 4 738 40035425
X59350.1 - 5 928

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
protein binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0