This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF777 SIRNA Assays ›
Gene Symbol:
ZNF777
Gene Name:
zinc finger protein 777
Gene Aliases:
-
Chromosome Location:
Chr.7: 149431363 - 149460962 on Build GRCh38
UniGene ID:
Hs.38512
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25915

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF777
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015694.2 NP_056509.2 5 1268
XM_005249980.3 XP_005250037.1 5 1254
XM_011516055.2 XP_011514357.1 5 1278
GenBank mRNA AB033111.1 - 5 1182
AK074515.1 - 3 154
AK075032.1 - 4 365
BC023985.2 - 4 332 2820826
BC105968.1 - 4 602

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0