This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ELP4 SIRNA Assays ›
Gene Symbol:
ELP4
Gene Name:
elongator acetyltransferase complex subunit 4
Gene Aliases:
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
Chromosome Location:
Chr.11: 31509729 - 31784525 on Build GRCh38
UniGene ID:
Hs.175534
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s25575

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ELP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288725.1 NP_001275654.1 1, 2 280
NM_001288726.1 NP_001275655.1 1, 2 280
NM_019040.4 NP_061913.3 1, 2 280
GenBank mRNA AK000505.1 - 1, 2 259
AK304885.1 - 1, 2 239
AL136677.1 - 1, 2 237
BC012514.1 - 1, 2 231 4509903
HY127287.1 - 1, 2 282

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
histone acetyltransferase activity
protein binding
phosphorylase kinase regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header