This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INTS7 SIRNA Assays ›
Gene Symbol:
INTS7
Gene Name:
integrator complex subunit 7
Gene Aliases:
C1orf73, INT7
Chromosome Location:
Chr.1: 211940328 - 212035660 on Build GRCh38
UniGene ID:
Hs.369285
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s24699

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: INTS7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199809.1 NP_001186738.1 18 2605
NM_001199811.1 NP_001186740.1 19 2710
NM_001199812.1 NP_001186741.1 19 2692
NM_015434.3 NP_056249.1 19 2752
NR_037667.1 20 2786
XM_011509396.2 XP_011507698.1 20 2745
GenBank mRNA AK001363.1 - 19 2552
AK001598.1 - 19 2634
AK022509.1 - 19 2612
AK022589.1 - 11 1419
AK297225.1 - 18 2484
AL117576.1 - 8 1026
AL133049.2 - 14 1921
BC020523.2 - 19 2586 3917030
BC030716.1 - 19 2607 3909231
BC033918.1 - 19 2611 3920829

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
snRNA processing
snRNA transcription from RNA polymerase II promoter
cellular response to ionizing radiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0