This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF345 SIRNA Assays ›
Gene Symbol:
ZNF345
Gene Name:
zinc finger protein 345
Gene Aliases:
HZF10
Chromosome Location:
Chr.19: 36850350 - 36893218 on Build GRCh38
UniGene ID:
Hs.362324
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s24618

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF345
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242472.1 NP_001229401.1 3 319
NM_001242474.1 NP_001229403.1 4 464
NM_001242475.1 NP_001229404.1 3 299
NM_001242476.1 NP_001229405.1 4 394
NM_003419.4 NP_003410.1 3 486
XM_011526704.2 XP_011525006.1 3 445
XM_017026568.1 XP_016882057.1 3 643
XM_017026569.1 XP_016882058.1 3 299
XM_017026570.1 XP_016882059.1 4 410
XM_017026571.1 XP_016882060.1 4 409
XM_017026572.1 XP_016882061.1 4 513
XM_017026573.1 XP_016882062.1 3 538
GenBank mRNA AK094146.1 - 2 893
AK297516.1 - 2 178
AK315069.1 - 3 319
BC032863.1 - 3 484 5270008
BP195158.1 - 4 462
DB028175.1 - 3 299
DB085650.1 - 4 386
X78933.1 - 3 304

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase III promoter
transcription from RNA polymerase II promoter
transcription from RNA polymerase III promoter
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0