This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB8A SIRNA Assays ›
Gene Symbol:
Rab8a
Gene Name:
RAB8A, member RAS oncogene family
Gene Aliases:
Mel, Rab8
Chromosome Location:
Chr.16: 19355047 - 19376776 on Build Rnor_6.0
UniGene ID:
Rn.16064
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s236213

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab8a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053998.2 NP_446450.2 2 145
GenBank mRNA BC071176.1 - 2 138
BC105863.1 - 2 145 7457017
JN019799.1 - 2 140

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_005370, BC002977, X56741, AK025165, BT007184, and others.
This siRNA also targets the following transcripts in Mouse: NM_023126, AK080740, AB232600, AK010862, S53270, and others.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle docking involved in exocytosis
axonogenesis
metabolic process
protein secretion
synaptic vesicle exocytosis
regulation of exocytosis
Rab protein signal transduction
cellular response to insulin stimulus
regulation of protein localization
cilium assembly
glucose homeostasis
positive regulation of glucose import
regulation of long-term neuronal synaptic plasticity
Golgi vesicle fusion to target membrane
regulation of protein transport
protein localization to plasma membrane
GTPase activity
GTP binding
GDP binding
protein kinase binding
myosin V binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0