This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPK11 SIRNA Assays ›
Gene Symbol:
Mapk11
Gene Name:
mitogen-activated protein kinase 11
Gene Aliases:
p38beta
Chromosome Location:
Chr.7: 130121678 - 130128633 on Build Rnor_6.0
UniGene ID:
Rn.45869
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s236086

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapk11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109532.2 NP_001103002.2 4 452
XM_008765792.1 XP_008764014.1 4 496
GenBank mRNA BC162032.1 - 5 488 9102947

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
protein phosphorylation
intracellular signal transduction
regulation of cardiac muscle cell proliferation
negative regulation of cardiac muscle cell proliferation
MAP kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0