This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GGPS1 SIRNA Assays ›
Gene Symbol:
Ggps1
Gene Name:
geranylgeranyl diphosphate synthase 1
Gene Aliases:
Crlf3, rGGPS1a, rGGPS1a1, rGGPS1a2, rGGPS1a3
Chromosome Location:
Chr.17: 53956185 - 53971039 on Build Rnor_6.0
UniGene ID:
Rn.54975
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s235972

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ggps1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007626.2 NP_001007627.1 4 887
XM_008771698.1 XP_008769920.1 4 905
XM_008771699.1 XP_008769921.1 3 812
XM_008771700.1 XP_008769922.1 3 834
GenBank mRNA AB118238.1 - 4 873
AB118239.1 - 4 873
AB118240.1 - 4 873
AB118241.1 - 4 873
AB118242.1 - 4 873
AB118243.1 - 4 873
BC090327.1 - 4 887 7125199

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

isoprenoid metabolic process
geranyl diphosphate biosynthetic process
geranylgeranyl diphosphate biosynthetic process
farnesyl diphosphate biosynthetic process
dimethylallyltranstransferase activity
farnesyltranstransferase activity
geranyltranstransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0