This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VPS9D1 SIRNA Assays ›
Gene Symbol:
Vps9d1
Gene Name:
VPS9 domain containing 1
Gene Aliases:
RGD1565149
Chromosome Location:
Chr.19: 56040527 - 56054185 on Build Rnor_6.0
UniGene ID:
Rn.204969
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s235640

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vps9d1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107440.2 NP_001100910.2 4 420
XM_006255767.1 XP_006255829.1 4 466
XM_006255768.2 XP_006255830.1 4 466
XM_008772613.1 XP_008770835.1 4 935
GenBank mRNA BC166767.1 - 4 420 7933213
FQ221344.1 - 4 431

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0