This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MBD2 SIRNA Assays ›
Gene Symbol:
Mbd2
Gene Name:
methyl-CpG binding domain protein 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 65814026 - 65885115 on Build Rnor_6.0
UniGene ID:
Rn.4085
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s235591

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mbd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115025.1 NP_001108497.1 4 892
GenBank mRNA BC158877.1 - 4 892 7934371

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010773, AF072243, BC046607, BC057289, BC026698, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
heart development
aging
response to mechanical stimulus
response to organic cyclic compound
positive regulation of Wnt signaling pathway
response to nutrient levels
response to estradiol
positive regulation of chromatin binding
regulation of cell proliferation
maternal behavior
ATP-dependent chromatin remodeling
cellular protein complex assembly
regulation of DNA methylation
embryonic organ development
cellular response to organic cyclic compound
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
DNA binding
chromatin binding
mRNA binding
methyl-CpG binding
protein domain specific binding
nucleosomal DNA binding
siRNA binding
C2H2 zinc finger domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0