This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB1B SIRNA Assays ›
Gene Symbol:
Rab1b LOC102555167
Gene Name:
RAB1B, member RAS oncogene family
ras-related protein Rab-1B-like
Gene Aliases:
Rab1bl
-
Chromosome Location:
Chr.1: 220483529 - 220491469 on Build Rnor_6.0
Chromosome Location:
Chr.1: 220480828 - 220482648 on Build Rnor_6.0
UniGene ID:
Rn.155100
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s235152

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab1b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109979.2 NP_001103449.1 240
GenBank mRNA BC085118.1 - 207
DN934711.1 - 258
EV762710.1 - 259
X13905.1 - 180
Gene Symbol: LOC102555167
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760197.1 XP_008758419.1 1 208

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
ER to Golgi vesicle-mediated transport
metabolic process
virion assembly
Rab protein signal transduction
positive regulation of glycoprotein metabolic process
regulation of autophagosome assembly
GTPase activity
GTP binding
GDP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0