This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC7A2 SIRNA Assays ›
Gene Symbol:
Slc7a2
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
Gene Aliases:
Cat2, Cat2a, Cat2b
Chromosome Location:
Chr.16: 54459409 - 54513349 on Build Rnor_6.0
UniGene ID:
Rn.48707
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s235013

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc7a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134686.2 NP_001128158.1 2 476
NM_022619.3 NP_072141.2 2 476
XM_008771281.1 XP_008769503.1 2 5555
XM_008771282.1 XP_008769504.1 3 435
XM_008771283.1 XP_008769505.1 2 5555
GenBank mRNA AF245001.1 - 2 477
AF245002.1 - 2 477

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007514, NM_001044740, DQ086834, AK171369, AK154621, and others.

Gene Ontology Categories:

Function(s) Process(es)

nitric oxide production involved in inflammatory response
amino acid transmembrane transport
nitric oxide biosynthetic process
arginine transport
macrophage activation
regulation of macrophage activation
regulation of inflammatory response
L-arginine import across plasma membrane
L-lysine import across plasma membrane
L-ornithine import across plasma membrane
cation transmembrane transport
L-arginine transport
L-alpha-amino acid transmembrane transport
L-ornithine transmembrane transport
L-ornithine transmembrane transporter activity
high-affinity arginine transmembrane transporter activity
high-affinity lysine transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
L-arginine transmembrane transporter activity
low-affinity L-arginine transmembrane transporter activity
high-affinity L-ornithine transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header