This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CREM SIRNA Assays ›
Gene Symbol:
Crem
Gene Name:
cAMP responsive element modulator
Gene Aliases:
Icer
Chromosome Location:
Chr.17: 57021704 - 57090888 on Build Rnor_6.0
UniGene ID:
Rn.10251
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s234984

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crem
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110860.2 NP_001104330.1 10 1776
NM_001271101.1 NP_001258030.1 10 1380
NM_001271102.1 NP_001258031.1 3 730
NM_001271245.1 NP_001258174.1 4 416
NM_001271246.1 NP_001258175.1 4 874
NM_001271247.1 NP_001258176.1 4 920
NM_001271248.1 NP_001258177.1 3 442
NM_017334.2 NP_059030.2 4 370
XM_006254049.2 XP_006254111.1 9 1734
XM_006254050.2 XP_006254112.1 9 1338
XM_006254051.2 XP_006254113.1 9 1560
XM_006254052.2 XP_006254114.1 8 1892
XM_006254053.2 XP_006254115.1 9 1514
XM_006254054.2 XP_006254116.1 8 1614
XM_006254055.2 XP_006254117.1 8 1478
XM_006254056.2 XP_006254118.1 8 1082
XM_006254057.2 XP_006254119.1 9 1581
XM_006254058.2 XP_006254120.1 8 1545
XM_006254059.2 XP_006254121.1 8 1149
XM_006254060.2 XP_006254122.1 7 1745
XM_006254061.2 XP_006254123.1 7 1349
XM_006254062.2 XP_006254124.1 6 1313
XM_006254063.2 XP_006254125.1 7 1464
XM_006254064.2 XP_006254126.1 8 1325
XM_006254065.2 XP_006254127.1 8 929
XM_006254066.2 XP_006254128.1 7 1289
XM_006254067.2 XP_006254129.1 7 893
XM_006254068.2 XP_006254130.1 5 1124
XM_006254069.2 XP_006254131.1 4 1153
XM_006254070.2 XP_006254132.1 3 1242
XM_008771691.1 XP_008769913.1 4 1199
XM_008771692.1 XP_008769914.1 4 795
XM_008771693.1 XP_008769915.1 3 1437
XM_008771694.1 XP_008769916.1 3 774
XM_008771695.1 XP_008769917.1 3 753
GenBank mRNA BC078899.1 - 10 1380 7133070
CO556486.1 - 3 391
FQ213544.1 - 3 730
FQ220921.1 - 3 729
S66024.1 - 4 367

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_182723, NM_001267570, NM_001267563, NM_182718, NM_183013, and others.
This siRNA also targets the following transcripts in Mouse: NM_001110859, NM_001110856, NM_013498, NM_001110853, NM_001271505, and others.

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
spermatogenesis
cAMP-mediated signaling
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0