siRNA Details
Gene Symbol: Vom2r67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099485.1 | NP_001092955.1 | 2 | 878 | ||
XM_008769941.1 | XP_008768163.1 | 4 | 1382 |

Gene Symbol: Vom2r67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099485.1 | NP_001092955.1 | 2 | 878 | ||
XM_008769941.1 | XP_008768163.1 | 4 | 1382 |