This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NUP160 SIRNA Assays ›
Gene Symbol:
NUP160
Gene Name:
nucleoporin 160
Gene Aliases:
-
Chromosome Location:
Chr.11: 47778083 - 47848544 on Build GRCh38
UniGene ID:
Hs.643526
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s23465

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NUP160
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318399.1 NP_001305328.1 2 406
NM_015231.2 NP_056046.1 2 406
NR_134636.1 2 406
GenBank mRNA AK026236.1 - 2 189
AK302396.1 - 2 192
AK304308.1 - 2 192
AL832029.1 - 2 196
BC009822.2 - 2 367
BC125227.1 - 2 329 40124592
BC125228.1 - 2 329 40124593
BI559779.1 - 2 417
BX647634.1 - 2 191
D83781.1 - 2 175

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
tRNA export from nucleus
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
protein transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
intracellular transport of virus
regulation of cellular response to heat
nucleocytoplasmic transporter activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0