This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASTN2 SIRNA Assays ›
Gene Symbol:
ASTN2
Gene Name:
astrotactin 2
Gene Aliases:
bA67K19.1
Chromosome Location:
Chr.9: 116425225 - 117415039 on Build GRCh38
UniGene ID:
Hs.601562
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s23381

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASTN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184734.1 NP_001171663.1 8 1319
NM_014010.4 NP_054729.3 22 3875
NM_198186.3 NP_937829.3 8 1640
NM_198187.3 NP_937830.3 8 1319
NM_198188.2 NP_937831.1 8 1319
GenBank mRNA AB014534.1 - 22 3875
AF116574.1 - 20 3096
AK024064.1 - 8 1290
AK127350.1 - 21 3096
AK315025.1 - 20 2939
BC018759.2 - 16 2324
BC029272.1 - 8 1640
BC093835.1 - 8 1153
BC101667.1 - 8 1152
BC143289.1 - 8 1153
BC143292.1 - 8 1153
BC143293.1 - 8 1153
BC146756.1 - 22 3875 9007192
BX649176.1 - 22 3210

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein localization to cell surface
calcium ion binding
inositol 1,3,4,5 tetrakisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0