This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NBEAL2 SIRNA Assays ›
Gene Symbol:
NBEAL2
Gene Name:
neurobeachin like 2
Gene Aliases:
BDPLT4, GPS
Chromosome Location:
Chr.3: 46979683 - 47009704 on Build GRCh38
UniGene ID:
Hs.437043
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s23314

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NBEAL2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015175.2 NP_055990.1 8 1023
XM_006713072.3 XP_006713135.1 8 1020
XM_011533533.2 XP_011531835.1 8 1020
XM_017006010.1 XP_016861499.1 8 1020
XM_017006011.1 XP_016861500.1 8 876
XM_017006012.1 XP_016861501.1 8 1020
XM_017006013.1 XP_016861502.1 8 1020
XM_017006014.1 XP_016861503.1 8 876
XM_017006016.1 XP_016861505.1 3 1597
GenBank mRNA AK092871.1 - 8 877
AK131104.1 - 8 863

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

platelet formation
phospholipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0