This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RHOX2A SIRNA Assays ›
Gene Symbol:
Rhox2a Rhox2b Rhox2c Rhox2e Rhox2f
Gene Name:
reproductive homeobox 2A
reproductive homeobox 2B
reproductive homeobox 2C
reproductive homeobox 2E
reproductive homeobox 2F
Gene Aliases:
4930539I12Rik, Rhox2, Rhox2.1
OTTMUSG00000017138, Rhox2.2, Rhox2a
OTTMUSG00000017149, Rhox2.3, Rhox2b
ENSMUSG00000063709, OTTMUSG00000017141, Rhox2.5, Rhox2d
EG434764, Rhox2.6, Rhox2e
Chromosome Location:
Chr.X: 37244402 - 37249690 on Build GRCm38
Chromosome Location:
Chr.X: 37411736 - 37416844 on Build GRCm38
Chromosome Location:
Chr.X: 37453480 - 37458417 on Build GRCm38
Chromosome Location:
Chr.X: 37530322 - 37541809 on Build GRCm38
Chromosome Location:
Chr.X: 37571421 - 37576159 on Build GRCm38
UniGene ID:
Mm.391240
Mm.415046
Mm.477362
Mm.455498
Mm.379498
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
siRNA ID s232839

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rhox2a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029203.2 NP_083479.1 2 185
GenBank mRNA AK015997.1 - 2 186
BC132216.1 - 2 179 40130493
BC132220.1 - 2 179 40130497
BC145534.1 - 2 179
DQ058639.1 - 2 186
Gene Symbol: Rhox2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099316.1 NP_001092786.1 2 163
GenBank mRNA BC049546.1 - 163
Gene Symbol: Rhox2c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099318.1 NP_001092788.1 185
Gene Symbol: Rhox2e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085348.1 NP_001078817.1 185
Gene Symbol: Rhox2f
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085356.1 NP_001078825.1 2 185

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0